Assay Details
| Gene Symbol: Ribc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007715.1 | NP_001007716.1 | 5 - 6 | 712 | 58 | |
| XM_006256799.2 | XP_006256861.1 | 4 - 5 | 899 | 58 | ||
| XM_006256800.2 | XP_006256862.1 | 5 - 6 | 852 | 58 | ||
| XM_006256801.2 | XP_006256863.1 | 5 - 6 | 863 | 58 | ||
| XM_006256802.2 | XP_006256864.1 | 5 - 6 | 767 | 58 | ||
| XM_006256803.2 | XP_006256865.1 | 5 - 6 | 770 | 58 | ||
| XM_006256804.2 | XP_006256866.1 | 5 - 6 | 752 | 58 | ||
| XM_006256805.2 | XP_006256867.1 | 5 - 6 | 748 | 58 | ||
| GenBank mRNA | BC078998.1 | - | 5 - 6 | 712 | 7112763 | 58 |
More Information
Updated Mapping Information
Set Membership: |
|

Ribc1
Genomic Map