Assay Details
| Gene Symbol: S100a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232805.2 | XP_006232867.1 | 1 - 2 | 102 | 79 | |
| GenBank mRNA | FQ223137.1 | - | 1 - 2 | 50 | 79 | |
| FQ226917.1 | - | 1 - 2 | 50 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the XM_006232805.2 transcripts.
This assay is in the 5'UTR region of the XM_006232805.2 transcripts.
Set Membership: |
|

S100a11
Genomic Map