Assay Details
| Gene Symbol: Tpd52l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044295.1 | NP_001037760.1 | 3 - 4 | 385 | 58 | |
| XM_006227737.2 | XP_006227799.1 | 3 - 4 | 478 | 58 | ||
| XM_006227739.2 | XP_006227801.1 | 3 - 4 | 478 | 58 | ||
| XM_008758674.1 | XP_008756896.1 | 3 - 4 | 477 | 58 | ||
| GenBank mRNA | BC099809.1 | - | 3 - 4 | 385 | 5621894 | 58 |
More Information
Updated Mapping Information
Set Membership: |
|

Tpd52l1
Genomic Map