Assay Details
| Gene Symbol: Dhrsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105914.1 | NP_001099384.1 | 3 - 4 | 275 | 63 | |
| XM_006248993.2 | XP_006249055.1 | 3 - 4 | 326 | 63 | ||
| XM_006248995.2 | XP_006249057.1 | 3 - 4 | 327 | 63 | ||
| XM_006248997.1 | XP_006249059.1 | 3 - 4 | 570 | 63 | ||
| GenBank mRNA | BC167053.1 | - | 3 - 4 | 254 | 7110020 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|

Dhrsx
Genomic Map