Assay Details
| Gene Symbol: Dhrsx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105914.1 | NP_001099384.1 | 1 - 2 | 97 | 97 | |
| XM_006248993.2 | XP_006249055.1 | 1 - 2 | 148 | 97 | ||
| XM_006248995.2 | XP_006249057.1 | 1 - 2 | 149 | 97 | ||
| GenBank mRNA | BC167053.1 | - | 1 - 2 | 76 | 7110020 | 97 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC167053.1, NM_001105914.1 transcripts.
Set Membership: |
|

Dhrsx
Genomic Map