Assay Details
| Gene Symbol: LOC100365773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271493.1 | NP_001258422.1 | 12 - 13 | 1380 | 102 | |
| GenBank mRNA | AB191343.1 | - | 12 - 13 | 1355 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100365773
Genomic Map