Assay Details
| Gene Symbol: LOC100365773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271493.1 | NP_001258422.1 | 6 - 7 | 790 | 75 | |
| GenBank mRNA | AB191343.1 | - | 6 - 7 | 765 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

LOC100365773
Genomic Map