Assay Details
| Gene Symbol: Jazf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006236536.2 | XP_006236598.1 | 1 - 2 | 2188 | 93 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006236536.2 transcripts.
Set Membership: |
|

Jazf1
Genomic Map