Assay Details
| Gene Symbol: Tril | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034010.1 | NP_001029182.1 | 1 | 82 | ||
| GenBank mRNA | BC100659.1 | - | 1 | 7935751 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tril
Genomic Map