Assay Details
| Gene Symbol: Npy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012614.2 | NP_036746.1 | 2 - 3 | 282 | 108 | |
| GenBank mRNA | JX157883.1 | - | 2 - 3 | 103 | 108 | |
| M15880.1 | - | 2 - 3 | 257 | 108 | ||
| M20373.1 | - | 2 - 3 | 237 | 108 | ||
| Gene Symbol: LOC100912228 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749745.3 | XP_003749793.1 | 295 | 108 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Npy
Genomic Map