Assay Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128083.1 | NP_001121555.1 | 2 - 3 | 1678 | 65 | |
| GenBank mRNA | CO393559.1 | - | 2 - 3 | 119 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim8
Genomic Map