Assay Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128083.1 | NP_001121555.1 | 4 - 5 | 1951 | 136 | |
| GenBank mRNA | CO393559.1 | - | 4 - 5 | 392 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trim8
Genomic Map