Assay Details
| Gene Symbol: Cnnm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107593.1 | NP_001101063.1 | 10 - 11 | 2007 | 61 | |
| XM_006231483.2 | XP_006231545.1 | 11 - 12 | 2969 | 61 | ||
| XM_008760426.1 | XP_008758648.1 | 10 - 11 | 2828 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P100783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9536 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600280 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733591 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125865 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683355 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20562 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7262 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8430 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P100783 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P100783 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P100783 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P910545 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P17282 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 53626 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35603 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10730280 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730281 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730278 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10730272 | - | - |
| Rat Genome 230 2.0 Array | 1376142_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381294_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1376142_at | - | - |
| Rat Genome 230 2.0 Array | 1390815_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390815_at | - | - |
| Rat Genome 230 2.0 Array | 1381294_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100029780.rn.1 | TC0100003657.rn.1 | RTA TC |
| Rat Expression Set 230 | 1376142_at | - | - |
| Rat Expression Set 230 | 1376142_PM_at | - | - |
| Rat Expression Set 230 | 1390815_at | - | - |
| Rat Expression Set 230 | 1390815_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10730272 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350425 | - | - |

Cnnm1
Genomic Map