Assay Details
| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139042.2 | NP_620611.2 | 16 - 17 | 2643 | 139 | |
| GenBank mRNA | AF024622.1 | - | 16 - 17 | 2643 | 139 | |
| U33847.1 | - | 736 | 139 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2g
Genomic Map