Assay Details
| Gene Symbol: Gucy2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139042.2 | NP_620611.2 | 17 - 18 | 2843 | 64 | |
| GenBank mRNA | AF024622.1 | - | 17 - 18 | 2843 | 64 | |
| U33847.1 | - | 936 | 64 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Gucy2g
Genomic Map