Assay Details
| Gene Symbol: Cnnm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011942.1 | NP_001011942.1 | 2 - 3 | 1873 | 103 | |
| XM_006231471.2 | XP_006231533.1 | 2 - 3 | 1966 | 103 | ||
| XM_008760413.1 | XP_008758635.1 | 2 - 3 | 1967 | 103 | ||
| GenBank mRNA | BC085930.1 | - | 2 - 3 | 1873 | 7110800 | 103 |
More Information
Updated Mapping Information
Set Membership: |
|

Cnnm2
Genomic Map