Assay Details
| Gene Symbol: Ldb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107601.1 | NP_001101071.1 | 6 - 7 | 1057 | 56 | |
| XM_006231493.1 | XP_006231555.1 | 6 - 7 | 889 | 56 | ||
| XM_006231494.1 | XP_006231556.1 | 6 - 7 | 889 | 56 | ||
| GenBank mRNA | BC166757.1 | - | 6 - 7 | 694 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ldb1
Genomic Map