Assay Details
| Gene Symbol: Wbp1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127484.1 | NP_001120956.1 | 3 - 4 | 452 | 120 | |
| XM_006231508.2 | XP_006231570.1 | 3 - 4 | 2447 | 120 | ||
| XM_008760435.1 | XP_008758657.1 | 3 - 4 | 1282 | 120 | ||
| XM_008760436.1 | XP_008758658.1 | 3 - 4 | 2559 | 120 | ||
| GenBank mRNA | BC166754.1 | - | 3 - 4 | 389 | 7929564 | 120 |
| FQ217301.1 | - | 3 - 4 | 697 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P294087 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P058577 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963077 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188662 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740580 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870251 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14570 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46738 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86644 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P294087 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058577 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P294087 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058577 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P058577 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P294087 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P294087 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P828898 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P129608 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 41758 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 42233 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47328 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10715867 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715866 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715863 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715865 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715868 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715864 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715869 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10715862 | - | - |
| Rat Genome 230 2.0 Array | 1372164_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1372164_at | - | - |
| Rat Genome 230 2.0 Array | 1375369_at | - | - |
| Rat Genome 230 2.0 Array | 1397681_at | - | - |
| Rat Genome 230 2.0 Array | 1397681_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375369_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1372164_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1397681_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375369_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375369_at | - | - |
| Affymetrix HT Rat Focus Array | 1397681_at | - | - |
| Affymetrix HT Rat Focus Array | 1372164_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100030618.rn.1 | TC0100003747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100030619.rn.1 | TC0100003747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100030620.rn.1 | TC0100003747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100030617.rn.1 | TC0100003747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100030621.rn.1 | TC0100003747.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100030616.rn.1 | TC0100003747.rn.1 | RTA TC |
| Rat Expression Set 230 | 1372164_PM_at | - | - |
| Rat Expression Set 230 | 1372164_at | - | - |
| Rat Expression Set 230 | 1375369_at | - | - |
| Rat Expression Set 230 | 1375369_PM_at | - | - |
| Rat Expression Set 230 | 1397681_PM_at | - | - |
| Rat Expression Set 230 | 1397681_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10715862 | - | - |
| Rat Genome U34 Set | rc_AI013397_at | - | - |
| Rat Genome U34 Set | rc_AA944906_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21394502 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364771 | - | - |

Wbp1l
Genomic Map