Assay Details
| Gene Symbol: Ackr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728033.3 | XP_002728079.3 | 1 - 2 | 1817 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002728033.3 transcripts.
Set Membership: |
|

Ackr1
Genomic Map