Assay Details
| Gene Symbol: Ptpn14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107200.1 | NP_001100670.1 | 16 - 17 | 3623 | 67 | |
| XM_006250453.2 | XP_006250515.1 | 17 - 18 | 3793 | 67 | ||
| XM_008769853.1 | XP_008768075.1 | 16 - 17 | 3411 | 67 | ||
| XM_008769854.1 | XP_008768076.1 | 17 - 18 | 3443 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
lymphangiogenesis
negative regulation of cell proliferation
peptidyl-tyrosine dephosphorylation
regulation of protein export from nucleus
regulation of nucleic acid-templated transcription
transcription cofactor activity
protein tyrosine phosphatase activity
receptor tyrosine kinase binding
lymphangiogenesis
negative regulation of cell proliferation
peptidyl-tyrosine dephosphorylation
regulation of protein export from nucleus
regulation of nucleic acid-templated transcription
transcription cofactor activity
protein tyrosine phosphatase activity
receptor tyrosine kinase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P027298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283135 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403506 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290356 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380326 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120739 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375902 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206036 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291136 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27229 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70351 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422314 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513884 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27282 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701515 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868121 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97378 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621522 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285639 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 918443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124037 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43094 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87283 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20798 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86147 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027298 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P027298 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P027298 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P23391 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54440 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80060 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68198 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101119 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34777 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10766693 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766679 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766683 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766695 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766682 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766692 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766700 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766690 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766687 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766684 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766691 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766681 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766694 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766697 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766699 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766696 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766680 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766685 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766678 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766698 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766686 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766688 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10766674 | - | - |
| Rat Genome 230 2.0 Array | 1383746_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383746_at | - | - |
| Rat Genome 230 2.0 Array | 1377427_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393670_at | - | - |
| Rat Genome 230 2.0 Array | 1374774_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1381891_at | - | - |
| Rat Genome 230 2.0 Array | 1393670_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1377427_at | - | - |
| Rat Genome 230 2.0 Array | 1381891_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374774_at | - | - |
| Affymetrix HT Rat Focus Array | 1383746_at | - | - |
| Affymetrix HT Rat Focus Array | 1393670_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383746_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393670_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1300015613.rn.1 | TC1300002197.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300015612.rn.1 | TC1300002197.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1300015611.rn.1 | TC1300002197.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374774_PM_at | - | - |
| Rat Expression Set 230 | 1377427_at | - | - |
| Rat Expression Set 230 | 1377427_PM_at | - | - |
| Rat Expression Set 230 | 1374774_at | - | - |
| Rat Expression Set 230 | 1383746_at | - | - |
| Rat Expression Set 230 | 1393670_PM_at | - | - |
| Rat Expression Set 230 | 1383746_PM_at | - | - |
| Rat Expression Set 230 | 1381891_at | - | - |
| Rat Expression Set 230 | 1381891_PM_at | - | - |
| Rat Expression Set 230 | 1393670_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10766674 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22014380 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359833 | - | - |

Ptpn14
Genomic Map