Assay Details
| Gene Symbol: Fh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017005.2 | NP_058701.2 | 4 - 5 | 562 | 80 | |
| GenBank mRNA | BC087598.1 | - | 4 - 5 | 562 | 7311498 | 80 |
| J04473.1 | - | 4 - 5 | 573 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fh
Genomic Map