Assay Details
| Gene Symbol: Fh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017005.2 | NP_058701.2 | 5 - 6 | 738 | 107 | |
| GenBank mRNA | BC087598.1 | - | 5 - 6 | 738 | 7311498 | 107 |
| FQ230216.1 | - | 1 - 2 | 83 | 107 | ||
| FQ232207.1 | - | 1 - 2 | 83 | 107 | ||
| J04473.1 | - | 5 - 6 | 749 | 107 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fh
Genomic Map