Assay Details
| Gene Symbol: Nhlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250280.1 | XP_006250342.1 | 1 - 2 | 301 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006250280.1 transcripts.
Set Membership: |
|

Nhlh1
Genomic Map