Assay Details
| Gene Symbol: Tp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_223012.8 | XP_223012.6 | 13 - 14 | 2946 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tp53bp2| Gene Symbol: Tp53bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_223012.8 | XP_223012.6 | 13 - 14 | 2946 | 56 | |
Set Membership: |
|