Assay Details
| Gene Symbol: Rbak | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006248867.1 | XP_006248929.1 | 2 - 3 | 315 | 63 | |
| XM_006248868.1 | XP_006248930.1 | 2 - 3 | 312 | 63 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006248867.1, XM_006248868.1 transcripts.
Set Membership: |
|

Rbak
Genomic Map