Assay Details
| Gene Symbol: Brat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751133.2 | XP_003751181.1 | 3 - 4 | 444 | 55 | |
| XM_006248968.2 | XP_006249030.1 | 3 - 4 | 399 | 55 | ||
| XM_006248972.1 | XP_006249034.1 | 4 - 5 | 560 | 55 | ||
More Information
Set Membership: |
|

Brat1
Genomic Map