Assay Details
| Gene Symbol: Tmprss11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003979.1 | NP_001003979.1 | 3 - 4 | 296 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss11c| Gene Symbol: Tmprss11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003979.1 | NP_001003979.1 | 3 - 4 | 296 | 134 | |
Set Membership: |
|