Assay Details
| Gene Symbol: Tmprss11c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003979.1 | NP_001003979.1 | 8 - 9 | 1099 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P454300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484635 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486915 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972569 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38887 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614412 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671643 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 224138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 133834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164263 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 146366 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P454300 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P454300 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P454300 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P454300 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5432 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93680 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35248 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 67040 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10772063 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772062 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772059 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772064 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772065 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10772055 | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400002689.rn.1 | TC1400000327.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10772055 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22393525 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349566 | - | - |

Tmprss11c
Genomic Map