Assay Details
| Gene Symbol: Tbc1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251004.2 | XP_006251066.1 | 10 - 11 | 1925 | 102 | |
| XM_006251005.2 | XP_006251067.1 | 10 - 11 | 1925 | 102 | ||
| XM_006251006.2 | XP_006251068.1 | 9 - 10 | 1319 | 102 | ||
| XM_006251007.1 | XP_006251069.1 | 9 - 10 | 1343 | 102 | ||
| XM_341215.7 | XP_341216.4 | 10 - 11 | 1925 | 102 | ||
More Information
Set Membership: |
|

Tbc1d1
Genomic Map