Assay Details
| Gene Symbol: Tbc1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006251004.2 | XP_006251066.1 | 18 - 19 | 3374 | 93 | |
| XM_006251005.2 | XP_006251067.1 | 17 - 18 | 3209 | 93 | ||
| XM_006251006.2 | XP_006251068.1 | 17 - 18 | 2768 | 93 | ||
| XM_006251007.1 | XP_006251069.1 | 17 - 18 | 2792 | 93 | ||
| XM_341215.7 | XP_341216.4 | 16 - 17 | 3089 | 93 | ||
More Information
Set Membership: |
|

Tbc1d1
Genomic Map