Assay Details
| Gene Symbol: Lnx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108358.1 | NP_001101828.1 | 9 - 10 | 2076 | 90 | |
| XM_006250902.2 | XP_006250964.1 | 9 - 10 | 2070 | 90 | ||
| XM_006250903.2 | XP_006250965.1 | 8 - 9 | 4108 | 90 | ||
| XM_006250904.2 | XP_006250966.1 | 8 - 9 | 4078 | 90 | ||
| GenBank mRNA | EU219404.1 | - | 8 - 9 | 1919 | 90 | |
| EU219405.1 | - | 8 - 9 | 1919 | 90 | ||
| EU219406.1 | - | 8 - 9 | 1787 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P236407 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36966 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809179 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601055 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867654 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622199 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 570841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442847 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480634 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 339029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8574 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494544 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796691 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424303 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743260 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727285 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153596 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 598021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939834 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 509657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195518 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70776 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50493 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156192 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38633 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236407 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236407 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P236407 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P236407 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P306784 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51241 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 47055 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100241 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48076 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10772288 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772292 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772282 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772291 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772283 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772284 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772289 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772285 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772293 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772290 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772286 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10772281 | - | - |
| Rat Genome 230 2.0 Array | 1385497_x_at | - | - |
| Rat Genome 230 2.0 Array | 1389464_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389464_at | - | - |
| Rat Genome 230 2.0 Array | 1385497_PM_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1389464_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1385497_x_at | - | - |
| Affymetrix HT Rat Focus Array | 1389464_at | - | - |
| Affymetrix HT Rat Focus Array | 1385497_PM_x_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400003269.rn.1 | TC1400000424.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400003270.rn.1 | TC1400000424.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389464_at | - | - |
| Rat Expression Set 230 | 1389464_PM_at | - | - |
| Rat Expression Set 230 | 1385497_x_at | - | - |
| Rat Expression Set 230 | 1385497_PM_x_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772281 | - | - |
| Rat Genome U34 Set | rc_AI112291_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20715479 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351112 | - | - |

Lnx1
Genomic Map