Assay Details
| Gene Symbol: Tmprss11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004020.1 | NP_001004020.1 | 8 - 9 | 965 | 154 | |
| XM_008770049.1 | XP_008768271.1 | 8 - 9 | 1059 | 154 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss11b
Genomic Map