Assay Details
| Gene Symbol: Gabra2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135779.1 | NP_001129251.1 | 7 - 8 | 741 | 123 | |
| XM_006250931.2 | XP_006250993.1 | 7 - 8 | 805 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of neurotransmitter levels
chloride transport
neurotransmitter transport
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
ion transmembrane transport
chloride transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
regulation of neurotransmitter levels
chloride transport
neurotransmitter transport
gamma-aminobutyric acid signaling pathway
chemical synaptic transmission
ion transmembrane transport
chloride transmembrane transport
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P378252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167560 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761566 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378683 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320470 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672312 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246864 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377164 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325148 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282226 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460439 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749845 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943043 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507705 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648552 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717725 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487660 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614898 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647065 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665327 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9771 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100376 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14379 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23521 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 108767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18609 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145586 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33235 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P378252 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P378252 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P378252 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P378252 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10772538 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772541 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772542 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772535 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772540 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772543 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772539 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772536 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772537 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10772534 | - | - |
| Rat Genome 230 2.0 Array | 1397614_at | - | - |
| Rat Genome 230 2.0 Array | 1397614_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400003721.rn.1 | TC1400000468.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400003723.rn.1 | TC1400000468.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400003722.rn.1 | TC1400000468.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400003716.rn.1 | TC1400000468.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400003715.rn.1 | TC1400000468.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400003720.rn.1 | TC1400000468.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400003714.rn.1 | TC1400000468.rn.1 | RTA TC |
| Rat Expression Set 230 | 1397614_at | - | - |
| Rat Expression Set 230 | 1397614_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772534 | - | - |
| Rat Genome U34 Set | L08491cds_at | - | - |
| Rat Genome U34 Set | L08491cds_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21976745 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352358 | - | - |

Gabra2
Genomic Map