Assay Details
| Gene Symbol: Paics | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080910.3 | NP_543186.1 | 4 - 5 | 479 | 119 | |
| XM_006250852.2 | XP_006250914.1 | 4 - 5 | 527 | 119 | ||
| XM_006250853.2 | XP_006250915.1 | 4 - 5 | 498 | 119 | ||
| XM_006250854.2 | XP_006250916.1 | 4 - 5 | 528 | 119 | ||
| GenBank mRNA | BC072508.1 | - | 4 - 5 | 469 | 7103198 | 119 |
| BC085711.1 | - | 4 - 5 | 479 | 7192781 | 119 | |
| D37979.1 | - | 2 - 3 | 384 | 119 | ||
| Gene Symbol: LOC100910308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003751352.3 | XP_003751400.2 | 1 - 2 | 255 | 119 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Paics
Genomic Map