Assay Details
| Gene Symbol: Cxcl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053647.1 | NP_446099.1 | 1 - 2 | 149 | 57 | |
| GenBank mRNA | AB060092.1 | - | 1 - 2 | 149 | 57 | |
| S77604.1 | - | 1 - 2 | 104 | 57 | ||
| U45965.1 | - | 1 - 2 | 112 | 57 | ||
| X65647.1 | - | 1 - 2 | 89 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cxcl2
Genomic Map