Assay Details
| Gene Symbol: Sept11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107208.2 | NP_001100678.1 | 342 | 69 | ||
| GenBank mRNA | BC167769.1 | - | 473 | 7456582 | 69 | |
| EU711414.1 | - | 342 | 69 | |||
| EU711415.1 | - | 342 | 69 | |||
| EU711416.1 | - | 342 | 69 | |||
| EU711417.1 | - | 342 | 69 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Sept11
Genomic Map