Assay Details
| Gene Symbol: Cnst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006250411.2 | XP_006250473.1 | 6 - 7 | 1731 | 86 | |
| XM_006250412.2 | XP_006250474.1 | 6 - 7 | 1120 | 86 | ||
| XM_006250413.2 | XP_006250475.1 | 6 - 7 | 1120 | 86 | ||
| XM_006250414.2 | XP_006250476.1 | 845 | 86 | |||
| XM_573525.5 | XP_573525.5 | 6 - 7 | 1120 | 86 | ||
More Information
Set Membership: |
|

Cnst
Genomic Map