Assay Details
| Gene Symbol: LOC498316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109085.1 | NP_001102555.1 | 2 - 3 | 374 | 88 | |
| XM_006250489.2 | XP_006250551.1 | 2 - 3 | 434 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC498316
Genomic Map