Assay Details
| Gene Symbol: Gemin8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007756.1 | NP_001007757.1 | 2 - 3 | 127 | 121 | |
| GenBank mRNA | BC079160.1 | - | 2 - 3 | 127 | 7129380 | 121 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC079160.1,NM_001007756.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gemin8
Genomic Map