Assay Details
| Gene Symbol: Ap1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127531.2 | NP_001121003.1 | 1 - 2 | 123 | 114 | |
| XM_006256860.2 | XP_006256922.1 | 1 - 2 | 195 | 114 | ||
| XM_006256862.2 | XP_006256924.1 | 1 - 2 | 194 | 114 | ||
| XM_008773167.1 | XP_008771389.1 | 1 - 2 | 195 | 114 | ||
| GenBank mRNA | BC166480.1 | - | 1 - 2 | 123 | 9102341 | 114 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC166480.1, NM_001127531.2, XM_006256860.2, XM_006256862.2, XM_008773167.1 transcripts.
Set Membership: |
|

Ap1s2
Genomic Map