Assay Details
| Gene Symbol: Ap1s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127531.2 | NP_001121003.1 | 3 - 4 | 406 | 153 | |
| XM_006256860.2 | XP_006256922.1 | 3 - 4 | 478 | 153 | ||
| XM_006256862.2 | XP_006256924.1 | 3 - 4 | 477 | 153 | ||
| XM_008773167.1 | XP_008771389.1 | 3 - 4 | 478 | 153 | ||
| GenBank mRNA | BC166480.1 | - | 3 - 4 | 406 | 9102341 | 153 |
More Information
Updated Mapping Information
Set Membership: |
|

Ap1s2
Genomic Map