Assay Details
| Gene Symbol: Uqcrh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009480.1 | NP_001009480.1 | 2 - 3 | 95 | 89 | |
| GenBank mRNA | BC086954.1 | - | 2 - 3 | 95 | 7368408 | 89 |
| FQ217127.1 | - | 2 - 3 | 140 | 89 | ||
| FQ217158.1 | - | 2 - 3 | 135 | 89 | ||
| FQ217230.1 | - | 2 - 3 | 132 | 89 | ||
| FQ217709.1 | - | 2 - 3 | 139 | 89 | ||
| FQ217710.1 | - | 2 - 3 | 125 | 89 | ||
| FQ217725.1 | - | 2 - 3 | 136 | 89 | ||
| FQ218028.1 | - | 2 - 3 | 125 | 89 | ||
| FQ221521.1 | - | 2 - 3 | 139 | 89 | ||
| FQ221648.1 | - | 2 - 3 | 139 | 89 | ||
| FQ221867.1 | - | 2 - 3 | 128 | 89 | ||
| FQ223801.1 | - | 2 - 3 | 125 | 89 | ||
| FQ223893.1 | - | 2 - 3 | 139 | 89 | ||
| FQ223943.1 | - | 2 - 3 | 136 | 89 | ||
| FQ224007.1 | - | 2 - 3 | 137 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Uqcrh
Genomic Map