Assay Details
| Gene Symbol: Slc6a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053818.2 | NP_446270.1 | 5 - 6 | 774 | 91 | |
| XM_006238625.2 | XP_006238687.1 | 4 - 5 | 780 | 91 | ||
| GenBank mRNA | BC128732.1 | - | 5 - 6 | 774 | 8364347 | 91 |
| L13600.1 | - | 4 - 5 | 671 | 91 | ||
| M88595.1 | - | 4 - 5 | 662 | 91 | ||
| M95413.1 | - | 5 - 6 | 725 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transmembrane transport
neurotransmitter transport
glycine transport
cation transmembrane transport
L-alpha-amino acid transmembrane transport
sodium:amino acid symporter activity
neurotransmitter:sodium symporter activity
protein binding
glycine transmembrane transporter activity
amino acid transmembrane transport
neurotransmitter transport
glycine transport
cation transmembrane transport
L-alpha-amino acid transmembrane transport
sodium:amino acid symporter activity
neurotransmitter:sodium symporter activity
protein binding
glycine transmembrane transporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P059525 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P034551 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146113 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286272 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73078 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262128 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833869 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118406 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12352 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941873 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26673 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059525 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034551 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059525 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034551 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P034551 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P059525 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P393273 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700254979 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 13907 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58155 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700254979 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10871461 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871452 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871463 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871462 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871458 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871454 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871456 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871445 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871457 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871450 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871455 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871460 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871451 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871459 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871446 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871453 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10871444 | - | - |
| Rat Genome 230 2.0 Array | 1373787_at | - | - |
| Rat Genome 230 2.0 Array | 1387693_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1369772_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1373787_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387693_a_at | - | - |
| Rat Genome 230 2.0 Array | 1369772_at | - | - |
| Affymetrix HT Rat Focus Array | 1373787_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1373787_at | - | - |
| Affymetrix HT Rat Focus Array | 1369772_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387693_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1387693_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1369772_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0500009144.rn.1 | TC0500001304.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0500009146.rn.1 | TC0500001304.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387693_a_at | - | - |
| Rat Expression Set 230 | 1373787_at | - | - |
| Rat Expression Set 230 | 1387693_PM_a_at | - | - |
| Rat Expression Set 230 | 1369772_PM_at | - | - |
| Rat Expression Set 230 | 1373787_PM_at | - | - |
| Rat Expression Set 230 | 1369772_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10871444 | - | - |
| Rat Genome U34 Set | U28975mRNA#2_at | - | - |
| Rat Genome U34 Set | U28975mRNA#2_g_at | - | - |
| Rat Genome U34 Set | L13600_at | - | - |
| Rat Genome U34 Set | rc_AA943735_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21070141 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357015 | - | - |
| RatRef-12 Expression BeadChip | ILMN_68859 | - | - |

Slc6a9
Genomic Map