Assay Details
| Gene Symbol: Bsnd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138979.2 | NP_620435.1 | 2 - 3 | 475 | 93 | |
| GenBank mRNA | AJ421029.1 | - | 2 - 3 | 506 | 93 | |
| BC081725.1 | - | 2 - 3 | 475 | 7127281 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bsnd
Genomic Map