Assay Details
| Gene Symbol: Dph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015007.1 | NP_001015007.1 | 1 - 2 | 235 | 71 | |
| GenBank mRNA | BC092656.1 | - | 1 - 2 | 235 | 7320904 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Dph2
Genomic Map