Assay Details
| Gene Symbol: Mpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006238749.2 | XP_006238811.1 | 6 - 7 | 873 | 77 | |
| XM_008764088.1 | XP_008762310.1 | 2026 | 77 | |||
| XM_008764089.1 | XP_008762311.1 | 1846 | 77 | |||
| XM_008764090.1 | XP_008762312.1 | 6 - 7 | 1060 | 77 | ||
| XM_345572.8 | XP_345573.5 | 6 - 7 | 1054 | 77 | ||
| GenBank mRNA | DQ013343.1 | - | 1 - 2 | 53 | 77 | |
| DQ013344.1 | - | 1 - 2 | 53 | 77 | ||
| DQ013345.1 | - | 1 - 2 | 53 | 77 | ||
More Information
Set Membership: |
|

Mpl
Genomic Map