Assay Details
| Gene Symbol: Mfrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108137.2 | NP_001101607.1 | 3 - 4 | 419 | 96 | |
| XM_006242933.2 | XP_006242995.1 | 3 - 4 | 864 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfrp
Genomic Map