Assay Details
| Gene Symbol: Usp28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108144.1 | NP_001101614.1 | 9 - 10 | 908 | 68 | |
| XM_006243006.2 | XP_006243068.1 | 9 - 10 | 1017 | 68 | ||
| XM_006243007.2 | XP_006243069.1 | 9 - 10 | 1014 | 68 | ||
| XM_006243008.2 | XP_006243070.1 | 9 - 10 | 1017 | 68 | ||
| XM_006243009.2 | XP_006243071.1 | 9 - 10 | 1017 | 68 | ||
| XM_006243010.2 | XP_006243072.1 | 9 - 10 | 1014 | 68 | ||
| XM_006243011.2 | XP_006243073.1 | 9 - 10 | 1017 | 68 | ||
| XM_006243012.2 | XP_006243074.1 | 9 - 10 | 1014 | 68 | ||
| GenBank mRNA | AB240643.1 | - | 9 - 10 | 917 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Usp28
Genomic Map