Assay Details
| Gene Symbol: Chrnb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_052806.2 | NP_434693.1 | 3 - 4 | 243 | 107 | |
| GenBank mRNA | AY574260.1 | - | 3 - 4 | 243 | 107 | |
| U42976.1 | - | 303 | 107 | |||
| X15834.1 | - | 355 | 107 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Chrnb4
Genomic Map