Assay Details
| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053642.2 | NP_446094.1 | 2 - 3 | 117 | 75 | |
| GenBank mRNA | AB052846.1 | - | 1 - 2 | 95 | 75 | |
| BC081704.1 | - | 2 - 3 | 117 | 7131154 | 75 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AB052846.1, BC081704.1, NM_053642.2 transcripts.
Set Membership: |
|

Sc5d
Genomic Map